Search
Search

Sult5a1| Gene Symbol: Sult5a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020564.3 | NP_065589.2 | 2 - 3 | 247 | 83 | |
| GenBank mRNA | AF026074.1 | - | 2 - 3 | 235 | 83 | |
| AK142243.1 | - | 2 - 3 | 248 | 83 | ||
| BC012677.1 | - | 2 - 3 | 243 | 4187578 | 83 | |
| CJ065192.1 | - | 2 - 3 | 248 | 83 | ||
Set Membership: |
|