Search
Search

Defb28| Gene Symbol: Defb28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037502.2 | NP_001032591.1 | 2 - 3 | 265 | 66 | |
| GenBank mRNA | AK131731.1 | - | 2 - 3 | 266 | 66 | |
| DQ012039.1 | - | 2 - 3 | 265 | 66 | ||
Set Membership: |
|