Search
Search

POFUT2| Gene Symbol: POFUT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961143.1 | XP_027816944.1 | 3 - 4 | 549 | 63 | |
| XM_027961146.1 | XP_027816947.1 | 3 - 4 | 525 | 63 | ||
Set Membership: |
|