Search
Search

TPD52L2| Gene Symbol: TPD52L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015099773.2 | XP_014955259.1 | 8 - 9 | 729 | 78 | |
| XM_015099774.2 | XP_014955260.1 | 7 - 8 | 667 | 78 | ||
| XM_015099775.2 | XP_014955261.1 | 6 - 7 | 685 | 78 | ||
| XM_015099776.2 | XP_014955262.1 | 5 - 6 | 596 | 78 | ||
Set Membership: |
|