Search
Search

ACIN1| Gene Symbol: ACIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004010335.4 | XP_004010384.3 | 18 - 19 | 3686 | 55 | |
| XM_027971629.1 | XP_027827430.1 | 18 - 19 | 3668 | 55 | ||
| XM_027971630.1 | XP_027827431.1 | 18 - 19 | 3635 | 55 | ||
| XM_027971631.1 | XP_027827432.1 | 18 - 19 | 3632 | 55 | ||
| XM_027971632.1 | XP_027827433.1 | 11 - 12 | 1553 | 55 | ||
| XM_027971633.1 | XP_027827434.1 | 11 - 12 | 1857 | 55 | ||
Set Membership: |
|