Search
Search

ACACA| Gene Symbol: ACACA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009256.1 | NP_001009256.1 | 22 - 23 | 3136 | 58 | |
| XM_027973950.1 | XP_027829751.1 | 21 - 22 | 3432 | 58 | ||
| XM_027973951.1 | XP_027829752.1 | 21 - 22 | 3122 | 58 | ||
| XM_027973952.1 | XP_027829753.1 | 20 - 21 | 3009 | 58 | ||
| GenBank mRNA | X80045.1 | - | 13 | 3136 | 58 | |
Set Membership: |
|