Search
Search

LRIG2| Gene Symbol: LRIG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015092177.2 | XP_014947663.2 | 11 - 12 | 2103 | 66 | |
| XM_027973238.1 | XP_027829039.1 | 9 - 10 | 1056 | 66 | ||
Set Membership: |
|