Search
Search

FAM13B| Gene Symbol: FAM13B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012178551.3 | XP_012033941.1 | 9 - 10 | 1351 | 71 | |
| XM_027970271.1 | XP_027826072.1 | 9 - 10 | 1278 | 71 | ||
| XM_027970272.1 | XP_027826073.1 | 9 - 10 | 1278 | 71 | ||
| XM_027970273.1 | XP_027826074.1 | 8 - 9 | 1182 | 71 | ||
| XM_027970274.1 | XP_027826075.1 | 9 - 10 | 1278 | 71 | ||
| XM_027970275.1 | XP_027826076.1 | 8 - 9 | 1182 | 71 | ||
Set Membership: |
|