Search
Search

PRKCE| Gene Symbol: PRKCE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004005978.3 | XP_004006027.1 | 14 - 15 | 2522 | 62 | |
| XM_027966880.1 | XP_027822681.1 | 14 - 15 | 2524 | 62 | ||
Set Membership: |
|