Search
Search

NR1H2| Gene Symbol: NR1H2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012190735.3 | XP_012046125.2 | 9 - 10 | 1500 | 75 | |
| XM_027977488.1 | XP_027833289.1 | 8 - 9 | 1454 | 75 | ||
| XM_027977489.1 | XP_027833290.1 | 9 - 10 | 1502 | 75 | ||
Set Membership: |
|