Search
Search

DENND1A| Gene Symbol: DENND1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027966460.1 | XP_027822261.1 | 9 - 10 | 827 | 63 | |
| XM_027966461.1 | XP_027822262.1 | 9 - 10 | 826 | 63 | ||
| XM_027966462.1 | XP_027822263.1 | 9 - 10 | 826 | 63 | ||
| XM_027966463.1 | XP_027822264.1 | 8 - 9 | 628 | 63 | ||
| XM_027966464.1 | XP_027822265.1 | 9 - 10 | 821 | 63 | ||
Set Membership: |
|