Search
Search
| Gene Symbol: TNIK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027958346.1 | XP_027814147.1 | 4 - 5 | 653 | 58 | |
| XM_027958349.1 | XP_027814150.1 | 4 - 5 | 653 | 58 | ||
| XM_027958352.1 | XP_027814153.1 | 4 - 5 | 653 | 58 | ||
| XM_027958358.1 | XP_027814159.1 | 4 - 5 | 654 | 58 | ||
| XM_027958366.1 | XP_027814167.1 | 4 - 5 | 653 | 58 | ||
| XM_027958371.1 | XP_027814172.1 | 4 - 5 | 657 | 58 | ||
| XM_027958380.1 | XP_027814181.1 | 4 - 5 | 654 | 58 | ||
| XM_027958384.1 | XP_027814185.1 | 4 - 5 | 653 | 58 | ||
| XM_027958388.1 | XP_027814189.1 | 4 - 5 | 653 | 58 | ||
Set Membership: |
|