Search
Search

TMCC2| Gene Symbol: TMCC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015099129.2 | XP_014954615.2 | 2 - 3 | 1099 | 79 | |
| XM_027976208.1 | XP_027832009.1 | 3 - 4 | 666 | 79 | ||
| XM_027976209.1 | XP_027832010.1 | 4 - 5 | 765 | 79 | ||