Search
Search

CNOT10| Gene Symbol: CNOT10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004018203.4 | XP_004018252.1 | 11 - 12 | 1531 | 64 | |
| XM_027957948.1 | XP_027813749.1 | 11 - 12 | 1565 | 64 | ||
Set Membership: |
|