Search
Search

METTL14| Gene Symbol: METTL14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004009592.4 | XP_004009641.1 | 2 - 3 | 328 | 98 | |
| XM_012179421.2 | XP_012034811.2 | 2 - 3 | 364 | 98 | ||
Set Membership: |
|