Search
Search

PRPF3| Gene Symbol: PRPF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002715596.3 | XP_002715642.1 | 11 - 12 | 1695 | 111 | |
| XM_017345910.1 | XP_017201399.1 | 11 - 12 | 1582 | 111 | ||
Set Membership: |
|