Search
Search

GTF3C2| Gene Symbol: GTF3C2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002709954.3 | XP_002710000.3 | 2 - 3 | 2236 | 90 | |
| XM_017340858.1 | XP_017196347.1 | 3 - 4 | 1436 | 90 | ||
| XM_017340859.1 | XP_017196348.1 | 3 - 4 | 799 | 90 | ||
Set Membership: |
|