Search
Search

RTCB| Gene Symbol: RTCB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711419.3 | XP_002711465.1 | 10 - 11 | 1378 | 63 | |
| XM_017342123.1 | XP_017197612.1 | 9 - 10 | 1512 | 63 | ||
Set Membership: |
|