Search
Search

KCND2| Gene Symbol: KCND2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082118.1 | NP_001075587.1 | 2 - 3 | 1277 | 66 | |
| GenBank mRNA | AF493547.1 | - | 2 - 3 | 1237 | 66 | |
| AF508735.1 | - | 2 - 3 | 1277 | 66 | ||
Set Membership: |
|