Search
Search

CELF1| Gene Symbol: CELF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008270634.2 | XP_008268856.1 | 7 - 8 | 736 | 90 | |
| XM_017346459.1 | XP_017201948.1 | 7 - 8 | 736 | 90 | ||
| XM_017346463.1 | XP_017201952.1 | 6 - 7 | 664 | 90 | ||
| XM_017346467.1 | XP_017201956.1 | 7 - 8 | 737 | 90 | ||
| XM_017346470.1 | XP_017201959.1 | 7 - 8 | 738 | 90 | ||
| XM_017346472.1 | XP_017201961.1 | 7 - 8 | 737 | 90 | ||
| XM_017346474.1 | XP_017201963.1 | 4 - 5 | 471 | 90 | ||
| XM_017346478.1 | XP_017201967.1 | 5 - 6 | 511 | 90 | ||
| XM_017346483.1 | XP_017201972.1 | 6 - 7 | 583 | 90 | ||
Set Membership: |
|