Search
Search

PSRC1| Gene Symbol: PSRC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002715807.3 | XP_002715853.2 | 3 - 4 | 336 | 57 | |
| XM_008264748.2 | XP_008262970.2 | 3 - 4 | 326 | 57 | ||
| XM_008264750.2 | XP_008262972.1 | 3 - 4 | 240 | 57 | ||
| XM_008264751.2 | XP_008262973.1 | 3 - 4 | 169 | 57 | ||
Set Membership: |
|