Search
Search

C5H16orf70| Gene Symbol: C5H16orf70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711619.3 | XP_002711665.1 | 9 - 10 | 829 | 73 | |
| XM_017342285.1 | XP_017197774.1 | 8 - 9 | 803 | 73 | ||
| XM_017342286.1 | XP_017197775.1 | 7 - 8 | 629 | 73 | ||
Set Membership: |
|