Search
Search

WDFY3| Gene Symbol: WDFY3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017347429.1 | XP_017202918.1 | 17 - 18 | 3049 | 55 | |
| XM_017347430.1 | XP_017202919.1 | 17 - 18 | 3049 | 55 | ||
| XM_017347431.1 | XP_017202920.1 | 17 - 18 | 3049 | 55 | ||
| XM_017347432.1 | XP_017202921.1 | 16 - 17 | 2995 | 55 | ||
| XM_017347433.1 | XP_017202922.1 | 17 - 18 | 3049 | 55 | ||
| XM_017347434.1 | XP_017202923.1 | 16 - 17 | 2965 | 55 | ||
| XM_017347435.1 | XP_017202924.1 | 13 - 14 | 2607 | 55 | ||
| XM_017347436.1 | XP_017202925.1 | 13 - 14 | 2607 | 55 | ||
Set Membership: |
|