Search
Search

TMEM59| Gene Symbol: TMEM59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002715081.3 | XP_002715127.3 | 1 - 2 | 268 | 60 | |
| XM_008265217.2 | XP_008263439.2 | 1 - 2 | 261 | 60 | ||
| XM_017346278.1 | XP_017201767.1 | 1 - 2 | 266 | 60 | ||
| XM_017346279.1 | XP_017201768.1 | 1 - 2 | 266 | 60 | ||
Set Membership: |
|