Search
Search

LRRN4| Gene Symbol: LRRN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008256295.2 | XP_008254517.1 | 4 - 5 | 1130 | 56 | |
| XM_017341767.1 | XP_017197256.1 | 3 - 4 | 1365 | 56 | ||
Set Membership: |
|