Search
Search

SLC39A11| Gene Symbol: SLC39A11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719500.3 | XP_002719546.1 | 8 - 9 | 885 | 59 | |
| XM_002719501.3 | XP_002719547.1 | 8 - 9 | 906 | 59 | ||
| XM_017349307.1 | XP_017204796.1 | 8 - 9 | 1005 | 59 | ||
| XM_017349308.1 | XP_017204797.1 | 8 - 9 | 984 | 59 | ||
Set Membership: |
|