Search
Search

NFE2L1| Gene Symbol: NFE2L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008271359.2 | XP_008269581.1 | 2 - 3 | 1112 | 68 | |
| XM_008271360.2 | XP_008269582.1 | 2 - 3 | 1100 | 68 | ||
| XM_008271361.2 | XP_008269583.1 | 2 - 3 | 1165 | 68 | ||
| XM_008271362.2 | XP_008269584.1 | 2 - 3 | 1126 | 68 | ||
| XM_017349171.1 | XP_017204660.1 | 2 - 3 | 1100 | 68 | ||
Set Membership: |
|