Search
Search

ENTPD4| Gene Symbol: ENTPD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002709315.3 | XP_002709361.1 | 11 - 12 | 1673 | 69 | |
| XM_008249546.2 | XP_008247768.1 | 11 - 12 | 1716 | 69 | ||
| XM_008249562.2 | XP_008247784.1 | 11 - 12 | 1692 | 69 | ||
| XM_017338071.1 | XP_017193560.1 | 11 - 12 | 1716 | 69 | ||
Set Membership: |
|