Search
Search

TRIM3| Gene Symbol: TRIM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008265270.2 | XP_008263492.1 | 7 - 8 | 1735 | 60 | |
| XM_008265282.2 | XP_008263504.1 | 5 - 6 | 1370 | 60 | ||
| XM_017344932.1 | XP_017200421.1 | 5 - 6 | 1303 | 60 | ||
Set Membership: |
|