Search
Search

DGKI| Gene Symbol: DGKI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017342646.1 | XP_017198135.1 | 3 - 4 | 522 | 71 | |
| XM_017342647.1 | XP_017198136.1 | 3 - 4 | 522 | 71 | ||
| XM_017342648.1 | XP_017198137.1 | 3 - 4 | 522 | 71 | ||
| XM_017342649.1 | XP_017198138.1 | 3 - 4 | 522 | 71 | ||
Set Membership: |
|