Search
Search

PDE10A| Gene Symbol: PDE10A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714982.3 | XP_002715028.1 | 6 - 7 | 666 | 58 | |
| XM_008263758.2 | XP_008261980.1 | 7 - 8 | 676 | 58 | ||
| XM_008263760.2 | XP_008261982.1 | 6 - 7 | 675 | 58 | ||
| XM_008263762.2 | XP_008261984.1 | 6 - 7 | 543 | 58 | ||
Set Membership: |
|