Search
Search

CDH19| Gene Symbol: CDH19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008261445.2 | XP_008259667.1 | 5 - 6 | 1059 | 102 | |
| XM_008261446.2 | XP_008259668.1 | 5 - 6 | 1093 | 102 | ||
Set Membership: |
|