Search
Search

FCRL4| Gene Symbol: FCRL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008264262.2 | XP_008262484.1 | 6 - 7 | 1338 | 67 | |
| XM_008264263.2 | XP_008262485.1 | 6 - 7 | 1334 | 67 | ||
| XM_008264264.2 | XP_008262486.1 | 6 - 7 | 1193 | 67 | ||
Set Membership: |
|