Search
Search

LARP4| Gene Symbol: LARP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008256452.2 | XP_008254674.1 | 7 - 8 | 1051 | 75 | |
| XM_008256453.2 | XP_008254675.1 | 6 - 7 | 960 | 75 | ||
| XM_008256456.2 | XP_008254678.1 | 6 - 7 | 626 | 75 | ||
| XM_017341854.1 | XP_017197343.1 | 7 - 8 | 1048 | 75 | ||
| XM_017341855.1 | XP_017197344.1 | 5 - 6 | 588 | 75 | ||
Set Membership: |
|