Search
Search

CCDC68| Gene Symbol: CCDC68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008261334.2 | XP_008259556.1 | 3 - 4 | 339 | 73 | |
| XM_008261335.2 | XP_008259557.1 | 3 - 4 | 304 | 73 | ||
| XM_017344193.1 | XP_017199682.1 | 4 - 5 | 469 | 73 | ||
Set Membership: |
|