Search
Search

ARMCX1| Gene Symbol: ARMCX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002720363.3 | XP_002720409.1 | 2 - 3 | 243 | 90 | |
| XM_017349652.1 | XP_017205141.1 | 2 - 3 | 243 | 90 | ||
Set Membership: |
|