Search
Search

OCIAD1| Gene Symbol: OCIAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017337643.1 | XP_017193132.1 | 1 - 2 | 59 | 92 | |
| XM_017337644.1 | XP_017193133.1 | 1 - 2 | 59 | 92 | ||
| XM_017337648.1 | XP_017193137.1 | 1 - 2 | 59 | 92 | ||
| XM_017337652.1 | XP_017193141.1 | 1 - 2 | 59 | 92 | ||
| XM_017337659.1 | XP_017193148.1 | 1 - 2 | 59 | 92 | ||
Set Membership: |
|