Search
Search

EPHA6| Gene Symbol: EPHA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008267078.1 | XP_008265300.1 | 14 - 15 | 4097 | 61 | |
| XM_017347166.1 | XP_017202655.1 | 9 - 10 | 1102 | 61 | ||
Set Membership: |
|