Search
Search

MDM4| Gene Symbol: MDM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002717573.3 | XP_002717619.1 | 10 - 11 | 2603 | 106 | |
| XM_008268530.2 | XP_008266752.1 | 9 - 10 | 2455 | 106 | ||
| XM_017347839.1 | XP_017203328.1 | 8 - 9 | 846 | 106 | ||
| XM_017347840.1 | XP_017203329.1 | 7 - 8 | 701 | 106 | ||
Set Membership: |
|