Search
Search

LOC4332360| Gene Symbol: LOC4332360 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775631.2 | XP_015631117.1 | 1 | 669 | 94 | |
| GenBank mRNA | AK119243.1 | - | 1 | 649 | 94 | |
| AK119717.1 | - | 1 | 649 | 94 | ||
| D12635.1 | - | 1 | 581 | 94 | ||
| EU846992.1 | - | 1 | 636 | 94 | ||
Set Membership: |
|