Search
Search

LOC4347829| Gene Symbol: LOC4347829 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756319.1 | XP_015611805.1 | 1 - 2 | 228 | 157 | |
| GenBank mRNA | AK071848.1 | - | 1 - 2 | 229 | 157 | |
| AK121320.1 | - | 1 - 2 | 227 | 157 | ||
Set Membership: |
|