Search
Search

LOC4337935| Gene Symbol: LOC4337935 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782477.2 | XP_015637963.1 | 1 - 2 | 510 | 107 | |
| GenBank mRNA | AK100216.1 | - | 1 - 2 | 374 | 107 | |
| AK103412.1 | - | 1 - 2 | 463 | 107 | ||
Set Membership: |
|