Search
Search

LOC4326396| Gene Symbol: LOC4326396 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785888.2 | XP_015641374.1 | 1 - 2 | 493 | 67 | |
| GenBank mRNA | AK110625.1 | - | 1 - 2 | 422 | 67 | |
| AY341842.1 | - | 1 - 2 | 444 | 67 | ||
| AY870605.1 | - | 1 - 2 | 408 | 67 | ||
Set Membership: |
|