Search
Search

LOC4324539| Gene Symbol: LOC4324539 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765569.2 | XP_015621055.1 | 8 - 9 | 2053 | 115 | |
| XM_015765570.2 | XP_015621056.1 | 8 - 9 | 2053 | 115 | ||
| XM_015765571.2 | XP_015621057.1 | 8 - 9 | 2053 | 115 | ||
| XM_015765572.2 | XP_015621058.1 | 7 - 8 | 1957 | 115 | ||
| XM_015765573.2 | XP_015621059.1 | 7 - 8 | 1903 | 115 | ||
| XM_015765574.2 | XP_015621060.1 | 8 - 9 | 2053 | 115 | ||
| XM_015765575.2 | XP_015621061.1 | 6 - 7 | 1807 | 115 | ||
| XM_015765576.2 | XP_015621062.1 | 7 - 8 | 1903 | 115 | ||
| XM_015765577.2 | XP_015621063.1 | 6 - 7 | 1807 | 115 | ||
| GenBank mRNA | AK062264.1 | - | 1 - 2 | 476 | 115 | |
Set Membership: |
|