Search
Search

LOC4344843| Gene Symbol: LOC4344843 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794717.2 | XP_015650203.1 | 2 | 850 | 157 | |
| GenBank mRNA | AF032976.1 | - | 2 | 832 | 157 | |
| AK061029.1 | - | 2 | 848 | 157 | ||
Set Membership: |
|