Search
Search

SSBP3| Gene Symbol: SSBP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261084.1 | NP_001248013.1 | 848 | 62 | ||
| XM_015139547.2 | XP_014995033.1 | 1166 | 62 | |||
| XM_015139553.2 | XP_014995039.1 | 1070 | 62 | |||
| XM_015139555.2 | XP_014995041.1 | 1016 | 62 | |||
| XM_028845282.1 | XP_028701115.1 | 1010 | 62 | |||
| XM_028845288.1 | XP_028701121.1 | 983 | 62 | |||
| GenBank mRNA | JU327905.1 | - | 663 | 62 | ||
| JU327906.1 | - | 663 | 62 | |||
| JU471112.1 | - | 743 | 62 | |||
| JU474989.1 | - | 743 | 62 | |||
| JU474990.1 | - | 824 | 62 | |||
| JV043592.1 | - | 848 | 62 | |||
| JV046624.1 | - | 767 | 62 | |||
| JV046625.1 | - | 848 | 62 | |||
| JV636069.1 | - | 726 | 62 | |||
| JV636070.1 | - | 807 | 62 | |||
| JV667785.1 | - | 738 | 62 | |||
Set Membership: |
|