Search
Search

CFAP70| Gene Symbol: CFAP70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015147383.2 | XP_015002869.1 | 3261 | 60 | ||
| XM_015147385.2 | XP_015002871.1 | 3379 | 60 | |||
| XM_015147386.2 | XP_015002872.1 | 3258 | 60 | |||
| XM_028826483.1 | XP_028682316.1 | 3051 | 60 | |||
| XM_028826484.1 | XP_028682317.1 | 3133 | 60 | |||
| XM_028826485.1 | XP_028682318.1 | 2914 | 60 | |||
| XM_028826487.1 | XP_028682320.1 | 2645 | 60 | |||
Set Membership: |
|