Search
Search

NR4A2| Gene Symbol: NR4A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266910.2 | NP_001253839.1 | 1758 | 71 | ||
| XM_015110132.2 | XP_014965618.1 | 1885 | 71 | |||
| XM_015110133.2 | XP_014965619.1 | 1856 | 71 | |||
| XM_015110134.2 | XP_014965620.1 | 1782 | 71 | |||
| GenBank mRNA | JU471922.1 | - | 1758 | 71 | ||
| JU471923.1 | - | 1758 | 71 | |||
Set Membership: |
|