Search
Search

IZUMO1| Gene Symbol: IZUMO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194623.1 | NP_001181552.1 | 401 | 67 | ||
| XM_015124385.2 | XP_014979871.1 | 1073 | 67 | |||
| XM_015124386.2 | XP_014979872.1 | 642 | 67 | |||
Set Membership: |
|