Search
Search

WDR47| Gene Symbol: WDR47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001088345.4 | XP_001088345.1 | 2777 | 81 | ||
| XM_002801688.3 | XP_002801734.1 | 2798 | 81 | |||
| XM_015146562.2 | XP_015002048.1 | 2678 | 81 | |||
| XM_015146565.2 | XP_015002051.1 | 2531 | 81 | |||
| XM_015146576.2 | XP_015002062.1 | 2657 | 81 | |||
| XM_015146583.2 | XP_015002069.1 | 2510 | 81 | |||
| XM_015146604.2 | XP_015002090.1 | 2774 | 81 | |||
| XM_015146610.2 | XP_015002096.1 | 2389 | 81 | |||
| XM_015146616.2 | XP_015002102.1 | 2368 | 81 | |||
| XM_028830808.1 | XP_028686641.1 | 2434 | 81 | |||
| XM_028830811.1 | XP_028686644.1 | 2413 | 81 | |||
| GenBank mRNA | JU333479.1 | - | 2748 | 81 | ||
| JU333480.1 | - | 2742 | 81 | |||
| JV709887.1 | - | 2701 | 81 | |||
Set Membership: |
|